Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192873.3 | NP_001179802.1 | 1 - 2 | 790 | 76 | |
GenBank mRNA | BT026251.1 | - | 1 - 2 | 724 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Bos taurus Probe spans exons |