Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163938.1 | NP_001157410.1 | 4 - 5 | 354 | 90 | |
GenBank mRNA | BT025354.1 | - | 4 - 5 | 331 | 90 | |
BT029841.1 | - | 4 - 5 | 339 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |