Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163938.1 | NP_001157410.1 | 6 - 7 | 780 | 66 | |
GenBank mRNA | BT025354.1 | - | 6 - 7 | 757 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |