Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163944.2 | NP_001157416.1 | 2 - 3 | 184 | 54 | |
GenBank mRNA | BC111275.1 | - | 2 - 3 | 185 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |