Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164024.1 | NP_001157496.1 | 31 - 32 | 4200 | 112 | |
GenBank mRNA | BC111679.1 | - | 2 - 3 | 163 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |