Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166518.3 | NP_001159990.3 | 2 - 3 | 151 | 63 | |
GenBank mRNA | AY057840.1 | - | 2 - 3 | 130 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |