Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206591.1 | NP_001193520.1 | 2 - 3 | 908 | 83 | |
GenBank mRNA | AB257750.1 | - | 1 - 2 | 90 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |