Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206597.1 | NP_001193526.1 | 8 - 9 | 1001 | 99 | |
GenBank mRNA | DV916511.1 | - | 6 - 7 | 691 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |