Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTBS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206600.1 | NP_001193529.1 | 6 - 7 | 984 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |