Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206610.1 | NP_001193539.1 | 3 - 4 | 427 | 57 | |
GenBank mRNA | BT025443.1 | - | 4 - 5 | 444 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |