Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206624.1 | NP_001193553.1 | 8 - 9 | 816 | 63 | |
GenBank mRNA | BC148945.1 | - | 8 - 9 | 816 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |