Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIFIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206700.1 | NP_001193629.1 | 3 - 4 | 256 | 75 | |
GenBank mRNA | EG707677.1 | - | 3 - 4 | 259 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |