Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206717.1 | NP_001193646.1 | 1 - 2 | 130 | 108 | |
GenBank mRNA | BC142047.1 | - | 1 - 2 | 127 | 108 | |
EV678475.1 | - | 1 - 2 | 130 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |