Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206717.1 | NP_001193646.1 | 2 - 3 | 236 | 67 | |
GenBank mRNA | BC142047.1 | - | 2 - 3 | 233 | 67 | |
EV678475.1 | - | 2 - 3 | 236 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |