Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206720.2 | NP_001193649.1 | 3 - 4 | 630 | 89 | |
GenBank mRNA | DV874786.1 | - | 3 - 4 | 630 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |