Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206784.2 | NP_001193713.1 | 1 - 2 | 417 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |