Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166562.1 | NP_001160034.1 | 2 - 3 | 1080 | 75 | |
GenBank mRNA | BC105227.1 | - | 2 - 3 | 708 | 75 | |
DV912573.1 | - | 2 - 3 | 703 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |