Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166562.1 | NP_001160034.1 | 3 - 4 | 1203 | 99 | |
GenBank mRNA | BC105227.1 | - | 3 - 4 | 831 | 99 | |
DV912573.1 | - | 3 - 4 | 826 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |