Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166564.1 | NP_001160036.1 | 4 - 5 | 838 | 90 | |
GenBank mRNA | BC142525.1 | - | 4 - 5 | 814 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |