Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167932.2 | NP_001161404.2 | 2 - 3 | 127 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |