Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC8B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177596.1 | NP_001171067.1 | 12 - 13 | 1721 | 90 | |
GenBank mRNA | BT026307.1 | - | 6 - 7 | 715 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |