Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190301.1 | NP_001177230.1 | 2 - 3 | 454 | 86 | |
GenBank mRNA | AB530321.1 | - | 2 - 3 | 454 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Bos taurus Probe spans exons |