Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191125.4 | NP_001178054.1 | 8 - 9 | 745 | 110 | |
GenBank mRNA | BC122812.1 | - | 8 - 9 | 762 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |