Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191136.3 | NP_001178065.1 | 4 - 5 | 543 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons |