Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGGBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191137.2 | NP_001178066.1 | 1 - 2 | 201 | 73 | |
GenBank mRNA | DV845525.1 | - | 1 - 2 | 201 | 73 | |
EW681465.1 | - | 1 - 2 | 200 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |