Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDYA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191147.2 | NP_001178076.2 | 3 - 4 | 399 | 71 | |
GenBank mRNA | BC109600.1 | - | 3 - 4 | 383 | 71 | |
DT833851.1 | - | 3 - 4 | 401 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |