Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNNM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191172.1 | NP_001178101.1 | 6 - 7 | 2347 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Bos taurus Probe spans exons |