Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191217.4 | NP_001178146.2 | 2 - 3 | 289 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |