Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191218.4 | NP_001178147.3 | 5 - 6 | 2613 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |