Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191219.3 | NP_001178148.1 | 3 - 4 | 404 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |