Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191219.3 | NP_001178148.1 | 8 - 9 | 1229 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |