Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191236.1 | NP_001178165.1 | 4 - 5 | 1235 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |