Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191246.5 | NP_001178175.1 | 2 - 3 | 276 | 90 | |
GenBank mRNA | BC148917.1 | - | 2 - 3 | 238 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |