Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206826.1 | NP_001193755.1 | 1 - 2 | 259 | 102 | |
NM_001206827.1 | NP_001193756.1 | 1 - 2 | 259 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |