Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206831.1 | NP_001193760.1 | 3 - 4 | 934 | 53 | |
GenBank mRNA | EH177301.1 | - | 3 - 4 | 417 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |