Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206852.1 | NP_001193781.1 | 1 - 2 | 393 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |