Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206853.1 | NP_001193782.1 | 6 - 7 | 1503 | 61 | |
GenBank mRNA | BC149309.1 | - | 4 - 5 | 769 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |