Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGLON5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206875.1 | NP_001193804.1 | 4 - 5 | 513 | 108 | |
GenBank mRNA | EE351204.1 | - | 2 - 3 | 165 | 108 | |
EH200051.1 | - | 4 - 5 | 490 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |