Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGLON5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206875.1 | NP_001193804.1 | 7 - 8 | 915 | 64 | |
GenBank mRNA | EE351204.1 | - | 5 - 6 | 567 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Bos taurus Probe spans exons |