Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206900.1 | NP_001193829.1 | 1 - 2 | 26 | 57 | |
GenBank mRNA | EV617278.1 | - | 1 - 2 | 30 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |