Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206900.1 | NP_001193829.1 | 3 - 4 | 848 | 54 | |
GenBank mRNA | EE377394.1 | - | 2 - 3 | 175 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |