Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206921.1 | NP_001193850.1 | 5 - 6 | 423 | 62 | |
NM_001206922.1 | NP_001193851.1 | 5 - 6 | 410 | 62 | ||
GenBank mRNA | CB448079.1 | - | 5 - 6 | 410 | 62 | |
DN533919.1 | - | 5 - 6 | 423 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |