Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206958.1 | NP_001193887.1 | 9 - 10 | 1629 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |