Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSTM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206964.1 | NP_001193893.1 | 1 - 2 | 93 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Bos taurus Probe spans exons |