Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206989.2 | NP_001193918.1 | 9 - 10 | 1751 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |