Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPACA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001207045.1 | NP_001193974.1 | 4 - 5 | 405 | 92 | |
GenBank mRNA | BC108171.1 | - | 4 - 5 | 406 | 92 | |
DT848094.1 | - | 4 - 5 | 375 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |