Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAMPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244141.1 | NP_001231070.1 | 9 - 10 | 1563 | 110 | |
GenBank mRNA | FE004250.1 | - | 1 - 2 | 139 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |