Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244181.1 | NP_001231110.1 | 9 - 10 | 1066 | 107 | |
NM_001244182.1 | NP_001231111.1 | 8 - 9 | 922 | 107 | ||
GenBank mRNA | BC105511.1 | - | 8 - 9 | 863 | 107 | |
DV889429.1 | - | 5 - 6 | 486 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |