Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF19A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191331.1 | NP_001178260.1 | 8 - 9 | 2027 | 75 | |
GenBank mRNA | BC151661.1 | - | 8 - 9 | 2006 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |