Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR3C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191349.2 | NP_001178278.1 | 3 - 4 | 2016 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |